Learn More
Biomatik Corporation Recombinant Human Filaggrin (FLG), partial, 100UG
Supplier:  Biomatik Corporation RPC24668
Encompass_Preferred
Recombinant Human Filaggrin (FLG), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: FLG. Target Synonyms: ATOD2; Epidermal filaggrin; FILA_HUMAN; Filaggrin; Filaggrin precursor; Fillagrin; FLG; Profilaggrin. Accession Number: P20930. Expression Region: 3838~4061aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 26.8kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5350