Learn More
Biomatik Corporation Recombinant Human Vitamin D-binding protein (GC), partial, 100UG
Supplier:  Biomatik Corporation RPC24674
Encompass_Preferred
Recombinant Human Vitamin D-binding protein (GC), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: GC. Target Synonyms: Gc protein-derived macrophage activating factor. Accession Number: P02774. Expression Region: 19~474aa. Tag Info: N-Terminal 6Xhis-Sumostar-Tagged. Theoretical MW: 67kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5351