Learn More
Biomatik Corporation Recombinant Bovine Glycolipid transfer protein (GLTP), 100UG
Supplier:  Biomatik Corporation RPC24685
Encompass_Preferred
Recombinant Bovine Glycolipid transfer protein (GLTP) is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Bovine (Bos taurus). Target Name: GLTP. Target Synonyms: GLTPGlycolipid transfer protein; GLTP. Accession Number: P68265. Expression Region: 2~209aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 25.8kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5355