Learn More
Biomatik Corporation Recombinant Sheep Interleukin-1 beta (IL1B), 100UG
Supplier:  Biomatik Corporation RPC24749
Encompass_Preferred
Recombinant Sheep Interleukin-1 beta (IL1B) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Sheep (Ovis aries). Target Name: IL1B. Target Synonyms: IL1BInterleukin-1 beta; IL-1 beta. Accession Number: P21621. Expression Region: 114~266aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 19.7kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5360