Learn More
Biomatik Corporation Recombinant Human Plastin-2 (LCP1), 100UG
Catalog No. 501505361
Encompass_Preferred
Recombinant Human Plastin-2 (LCP1) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: LCP1. Target Synonyms: L-plastin; LC64PLymphocyte cytosolic protein 1; LCP-1. Accession Number: P13796. Expression Region: 2~627aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 72.2kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5361 Supplier Biomatik Corporation Supplier No. RPC24785
Recombinant Human Plastin-2 (LCP1) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: LCP1. Target Synonyms: L-plastin; LC64PLymphocyte cytosolic protein 1; LCP-1. Accession Number: P13796. Expression Region: 2~627aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 72.2kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.