Learn More
Biomatik Corporation Recombinant Mouse Microtubule-associated protein tau (Mapt), 100UG
Catalog No. 501505362
Encompass_Preferred
Recombinant Mouse Microtubule-associated protein tau (Mapt) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Mouse (Mus musculus). Target Name: Mapt. Target Synonyms: Neurofibrillary tangle proteinPaired helical filament-tauPHF-tau. Accession Number: P10637. Expression Region: 2~733aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 78.1kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5362 Supplier Biomatik Corporation Supplier No. RPC24806
Recombinant Mouse Microtubule-associated protein tau (Mapt) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Mouse (Mus musculus). Target Name: Mapt. Target Synonyms: Neurofibrillary tangle proteinPaired helical filament-tauPHF-tau. Accession Number: P10637. Expression Region: 2~733aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 78.1kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.