Learn More
Biomatik Corporation Recombinant Human Protein-methionine sulfoxide oxidase MICAL2 (MICAL2), partial, 100UG
Catalog No. 501505365
Encompass_Preferred
Recombinant Human Protein-methionine sulfoxide oxidase MICAL2 (MICAL2), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: MICAL2. Target Synonyms: Molecule interacting with CasL protein 2; MICAL-2. Accession Number: O94851. Expression Region: 1~495aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 58.5kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5365 Supplier Biomatik Corporation Supplier No. RPC24815
Recombinant Human Protein-methionine sulfoxide oxidase MICAL2 (MICAL2), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: MICAL2. Target Synonyms: Molecule interacting with CasL protein 2; MICAL-2. Accession Number: O94851. Expression Region: 1~495aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 58.5kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.