Learn More
Biomatik Corporation Recombinant Human Neprilysin (MME), partial, 100UG
Supplier:  Biomatik Corporation RPC24818
Encompass_Preferred
Recombinant Human Neprilysin (MME), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: MME. Target Synonyms: AtriopeptidaseCommon acute lymphocytic leukemia antigen; CALLAEnkephalinaseNeutral endopeptidase 24.11; NEP; Neutral endopeptidase; Skin fibroblast elastase; SFE; CD10. Accession Number: P08473. Expression Region: 52~750aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 81.8kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5366