Learn More
Biomatik Corporation Recombinant Human Nucleolin (NCL), partial, 100UG
Catalog No. 501505368
Encompass_Preferred
Recombinant Human Nucleolin (NCL), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: NCL. Target Synonyms: C23; FLJ45706; MS1116; NCL; Nucl; NUCL_HUMAN; Nucleolin; Protein C23. Accession Number: P19338. Expression Region: 2~482aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 54.4kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5368 Supplier Biomatik Corporation Supplier No. RPC24841
Recombinant Human Nucleolin (NCL), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: NCL. Target Synonyms: C23; FLJ45706; MS1116; NCL; Nucl; NUCL_HUMAN; Nucleolin; Protein C23. Accession Number: P19338. Expression Region: 2~482aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 54.4kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.