Learn More
Biomatik Corporation Recombinant Rat Syndecan-1 (Sdc1), partial, 100UG
Catalog No. 501505376
Encompass_Preferred
Recombinant Rat Syndecan-1 (Sdc1), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Rat (Rattus norvegicus). Target Name: Sdc1. Target Synonyms: CD138. Accession Number: P26260. Expression Region: 23~257aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 26.5kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5376 Supplier Biomatik Corporation Supplier No. RPC24935
Recombinant Rat Syndecan-1 (Sdc1), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Rat (Rattus norvegicus). Target Name: Sdc1. Target Synonyms: CD138. Accession Number: P26260. Expression Region: 23~257aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 26.5kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.