Learn More
Biomatik Corporation Recombinant Human cytomegalovirus Enhanced green fluorescent protein (egfp), 100UG
Supplier:  Biomatik Corporation RPC25034
Encompass_Preferred
Recombinant Human cytomegalovirus Enhanced green fluorescent protein (egfp) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Human cytomegalovirus (HHV-5) (Human herpesvirus 5). Target Name: egfp. Accession Number: C5MKY7. Expression Region: 1~239aa. Tag Info: N-Terminal 10Xhis-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 30.9kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5382