Learn More
Biomatik Corporation Recombinant Mycoplasma hyopneumoniae 46KDA surface antigen (p46), 100UG
Supplier:  Biomatik Corporation RPC25073
Encompass_Preferred
Recombinant Mesomycoplasma hyopneumoniae 46 kDa surface antigen (p46) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Mesomycoplasma hyopneumoniae (strain 232). Target Name: p46. Target Synonyms: p46. Accession Number: P0C0J7. Expression Region: 28~416aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 44.5kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5386