Learn More
Biomatik Corporation Recombinant Cryptococcus neoformans var. neoformans serotype D Orotate phosphoribosyltransferase (URA5), 100UG
Catalog No. 501505387
Encompass_Preferred
Recombinant Cryptococcus neoformans var. neoformans serotype D Orotate phosphoribosyltransferase (URA5) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Cryptococcus neoformans var. neoformans serotype D (strain JEC21 / ATCC MYA-565) (Filobasidiella neoformans). Target Name: URA5. Target Synonyms: URA5; CNG03730; Orotate phosphoribosyltransferase; OPRT; OPRTase; EC 2.4.2.10. Accession Number: P0CS95. Expression Region: 1~225aa. Tag Info: N-Terminal 10Xhis-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 28.4kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5387 Supplier Biomatik Corporation Supplier No. RPC25081
Recombinant Cryptococcus neoformans var. neoformans serotype D Orotate phosphoribosyltransferase (URA5) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Cryptococcus neoformans var. neoformans serotype D (strain JEC21 / ATCC MYA-565) (Filobasidiella neoformans). Target Name: URA5. Target Synonyms: URA5; CNG03730; Orotate phosphoribosyltransferase; OPRT; OPRTase; EC 2.4.2.10. Accession Number: P0CS95. Expression Region: 1~225aa. Tag Info: N-Terminal 10Xhis-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 28.4kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.