Learn More
Biomatik Corporation Recombinant Mouse Lymphocyte antigen 6C1 (Ly6c1), 100UG
Supplier:  Biomatik Corporation RPC25084
Encompass_Preferred
Recombinant Mouse Lymphocyte antigen 6C1 (Ly6c1) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Mouse (Mus musculus). Target Name: Ly6c1. Target Synonyms: Ly6c1; Lymphocyte antigen 6C1; Ly-6C1. Accession Number: P0CW02. Expression Region: 27~109aa. Tag Info: N-Terminal 6Xhis-Sumostar-Tagged. Theoretical MW: 25.1kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5388