Learn More
Biomatik Corporation Recombinant Staphylococcus aureus Enterotoxin type G (entG), 100UG
Supplier:  Biomatik Corporation RPC20132
Encompass_Preferred
Recombinant Staphylococcus aureus Enterotoxin type G (entG) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Staphylococcus aureus (strain N315). Target Name: entG. Target Synonyms: SEG. Accession Number: P0A0L7. Expression Region: 26~258aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 43kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4615