Learn More
Biomatik Corporation Recombinant Rat Trypsin-4 (Try4), 100UG
Supplier:  Biomatik Corporation RPC25092
Encompass_Preferred
Recombinant Rat Trypsin-4 (Try4) is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Rat (Rattus norvegicus). Target Name: Try4. Target Synonyms: Pretrypsinogen IVTrypsin IV. Accession Number: P12788. Expression Region: 24~247aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 26.1kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5389