Learn More
Biomatik Corporation Recombinant Glycine max 2S albumin, 100UG
Supplier:  Biomatik Corporation RPC25129
Encompass_Preferred
Recombinant Glycine max 2S seed storage albumin protein is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Glycine max (Soybean) (Glycine hispida). Target Name: Glycine max 2S seed storage albumin protein. Target Synonyms: 2S albumin; GM2S-1; Napin-type 2S albumin 3; Cleaved into: 2S albumin small chain; Aspartic acid-rich peptide; Lunasin; 2S albumin large chain; 8 kDa methionine-rich protein; 8 kDa MRP. Accession Number: P19594. Expression Region: 22~158aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 18.2kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5393