Learn More
Biomatik Corporation Recombinant Bacillus methanolicus NAD-dependent methanol dehydrogenase (mdh), 100UG
Supplier:  Biomatik Corporation RPC25143
Encompass_Preferred
Recombinant Bacillus methanolicus NAD-dependent methanol dehydrogenase (mdh) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Bacillus methanolicus. Target Name: mdh. Target Synonyms: Type 3 alcohol dehydrogenase. Accession Number: P31005. Expression Region: 2~381aa. Tag Info: N-Terminal 10Xhis-Tagged. Theoretical MW: 42.4kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5394