Learn More
Biomatik Corporation Recombinant Gardnerella vaginalis 5-1 Modification methylase AccI (GV51_0914), 100UG
Supplier:  Biomatik Corporation RPC25153
Encompass_Preferred
Recombinant Gardnerella vaginalis 5-1 Site-specific DNA-methyltransferase (adenine-specific) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Gardnerella vaginalis 5-1. Target Name: Gardnerella vaginalis 5-1 Site-specific DNA-methyltransferase (adenine-specific). Accession Number: D6SZ78. Expression Region: 1~525aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 62.2kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5395