Learn More
Biomatik Corporation Recombinant Saccharomyces cerevisiae 2-deoxyglucose-6-phosphate phosphatase 1 (DOG1), 100UG
Supplier:  Biomatik Corporation RPC25171
Encompass_Preferred
Recombinant Saccharomyces cerevisiae 2-deoxyglucose-6-phosphate phosphatase 1 (DOG1) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Saccharomyces cerevisiae (strain ATCC 204508 / S288c) (Baker's yeast). Target Name: DOG1. Target Synonyms: DOG1; YHR044C2-deoxyglucose-6-phosphate phosphatase 1; 2-DOG-6-P 1; 2-deoxyglucose-6-phosphatase 1; EC 3.1.3.68. Accession Number: P38774. Expression Region: 1~246aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 29.1kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5398