Learn More
Biomatik Corporation Recombinant Escherichia coli Type 1 fimbrin D-mannose specific adhesin (fimH), 100UG
Catalog No. 501504616
Encompass_Preferred
Recombinant Escherichia coli Type 1 fimbrin D-mannose specific adhesin (fimH) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Escherichia coli (strain K12). Target Name: fimH. Target Synonyms: fimH; b4320; JW4283Type 1 fimbrin D-mannose specific adhesin; Protein FimH. Accession Number: P08191. Expression Region: 22~300aa. Tag Info: N-Terminal 6Xhis-Gst-Tagged. Theoretical MW: 59.1kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4616 Supplier Biomatik Corporation Supplier No. RPC20134
Recombinant Escherichia coli Type 1 fimbrin D-mannose specific adhesin (fimH) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Escherichia coli (strain K12). Target Name: fimH. Target Synonyms: fimH; b4320; JW4283Type 1 fimbrin D-mannose specific adhesin; Protein FimH. Accession Number: P08191. Expression Region: 22~300aa. Tag Info: N-Terminal 6Xhis-Gst-Tagged. Theoretical MW: 59.1kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.