Learn More
Biomatik Corporation Recombinant Hirudo nipponia Guamerin, 100UG
Catalog No. 501505401
Encompass_Preferred
Recombinant Hirudo nipponia Guamerin is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Hirudo nipponia (Korean blood-sucking leech). Target Name: Hirudo nipponia Guamerin. Target Synonyms: Guamerin. Accession Number: P46443. Expression Region: 1~57aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 8.1kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5401 Supplier Biomatik Corporation Supplier No. RPC25189
Recombinant Hirudo nipponia Guamerin is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Hirudo nipponia (Korean blood-sucking leech). Target Name: Hirudo nipponia Guamerin. Target Synonyms: Guamerin. Accession Number: P46443. Expression Region: 1~57aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 8.1kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.