Learn More
Biomatik Corporation Recombinant Anemonia sulcata Delta-actitoxin-Avd1c, 100UG
Catalog No. 501505402
Encompass_Preferred
Recombinant Anemonia sulcata Delta-actitoxin-Avd1c is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Anemonia sulcata (Mediterranean snakelocks sea anemone). Target Name: Anemonia sulcata Delta-actitoxin-Avd1c. Target Synonyms: ATX-II. Accession Number: P01528. Expression Region: 1~47aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 6.9kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5402 Supplier Biomatik Corporation Supplier No. RPC25214
Recombinant Anemonia sulcata Delta-actitoxin-Avd1c is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Anemonia sulcata (Mediterranean snakelocks sea anemone). Target Name: Anemonia sulcata Delta-actitoxin-Avd1c. Target Synonyms: ATX-II. Accession Number: P01528. Expression Region: 1~47aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 6.9kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.