Learn More
Biomatik Corporation Recombinant Staphylococcus aureus Phospholipase C (hlb), 100UG
Catalog No. 501505406
Encompass_Preferred
Recombinant Staphylococcus aureus Phospholipase C (hlb) is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Staphylococcus aureus (strain MRSA252). Target Name: hlb. Target Synonyms: Beta-hemolysinBeta-toxinSphingomyelinaseplc. Accession Number: P09978. Expression Region: 35~330aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 35.7kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5406 Supplier Biomatik Corporation Supplier No. RPC25237
Recombinant Staphylococcus aureus Phospholipase C (hlb) is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Staphylococcus aureus (strain MRSA252). Target Name: hlb. Target Synonyms: Beta-hemolysinBeta-toxinSphingomyelinaseplc. Accession Number: P09978. Expression Region: 35~330aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 35.7kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.