Learn More
Biomatik Corporation Recombinant Hepatitis delta virus genotype I Small delta antigen, 100UG
Supplier:  Biomatik Corporation RPC25267
Encompass_Preferred
Recombinant Hepatitis delta virus genotype I Small delta antigen is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Hepatitis delta virus genotype I (isolate Italian) (HDV). Target Name: Hepatitis delta virus genotype I Small delta antigen. Target Synonyms: p24. Accession Number: P06934. Expression Region: 1~195aa. Tag Info: N-Terminal 6Xhis-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 25.4kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5411