Learn More
Biomatik Corporation Recombinant Streptococcus agalactiae Toxin zeta, 100UG
Supplier:  Biomatik Corporation RPC25280
Encompass_Preferred
Recombinant Streptococcus agalactiae Toxin zeta is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Streptococcus agalactiae. Target Name: Streptococcus agalactiae Toxin zeta. Target Synonyms: Toxin zeta; UDP-N-acetylglucosamine kinase; UNAG kinase; EC 2.7.1.176. Accession Number: P0A4M2. Expression Region: 1~287aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 34.7kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5414