Learn More
Biomatik Corporation Recombinant Rotavirus X Outer capsid protein VP4, partial, 100UG
Catalog No. 501505416
Encompass_Preferred
Recombinant Rotavirus X Outer capsid protein VP4 is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Rotavirus X (isolate RVX/Human/Bangladesh/NADRV-B219/2002/GXP[X]) (RV ADRV-N) (Rotavirus (isolate novel adult diarrhea rotavirus-B219)). Target Name: Rotavirus X Outer capsid protein VP4. Target Synonyms: Hemagglutinin. Accession Number: A9Q1L0. Expression Region: 1~249aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 30.8kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5416 Supplier Biomatik Corporation Supplier No. RPC25310
Recombinant Rotavirus X Outer capsid protein VP4 is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Rotavirus X (isolate RVX/Human/Bangladesh/NADRV-B219/2002/GXP[X]) (RV ADRV-N) (Rotavirus (isolate novel adult diarrhea rotavirus-B219)). Target Name: Rotavirus X Outer capsid protein VP4. Target Synonyms: Hemagglutinin. Accession Number: A9Q1L0. Expression Region: 1~249aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 30.8kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.