Learn More
Biomatik Corporation Recombinant Saccharomyces cerevisiae Ubiquitin-like protein SMT3 (SMT3), 100UG
Supplier:  Biomatik Corporation RPC25357
Encompass_Preferred
Recombinant Saccharomyces cerevisiae Ubiquitin-like protein SMT3 (SMT3) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Saccharomyces cerevisiae (strain ATCC 204508 / S288c) (Baker's yeast). Target Name: SMT3. Target Synonyms: DmSUMO 1; Small Ubiquitin-like modifier; SMT3; SMT3_YEAST; Ubiquitin like protein of the SUMO family; Ubiquitin like protein SMT3; Ubiquitin-like protein SMT3. Accession Number: Q12306. Expression Region: 2~98aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 13.1kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5420