Learn More
Biomatik Corporation Recombinant Human Inactive tyrosine-protein kinase 7 (PTK7), partial, 100UG
Supplier:  Biomatik Corporation RPC25368
Encompass_Preferred
Recombinant Human Inactive tyrosine-protein kinase 7 (PTK7), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: PTK7. Target Synonyms: Colon carcinoma kinase 4; CCK-4Protein-tyrosine kinase 7Pseudo tyrosine kinase receptor 7Tyrosine-protein kinase-like 7. Accession Number: Q13308. Expression Region: 31~704aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 76.6kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5421