Learn More
Biomatik Corporation Recombinant Human Glucokinase regulatory protein (GCKR), 100UG
Catalog No. 501505422
Encompass_Preferred
Recombinant Human Glucokinase regulatory protein (GCKR) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: GCKR. Target Synonyms: FGQTL5; GCK; GCKR; GCKR_HUMAN; GKRP; GLRE; glucokinase (hexokinase 4) regulator; Glucokinase; Glucokinase regulator; Glucokinase regulatory protein; Hexokinase 4 regulator; Hexokinase type IV; Hexokinase-4; Hexokinase-D; HK IV; HK4; OTTHUMP00000158533. Accession Number: Q14397. Expression Region: 1~625aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 70.7kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5422 Supplier Biomatik Corporation Supplier No. RPC25369
Recombinant Human Glucokinase regulatory protein (GCKR) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: GCKR. Target Synonyms: FGQTL5; GCK; GCKR; GCKR_HUMAN; GKRP; GLRE; glucokinase (hexokinase 4) regulator; Glucokinase; Glucokinase regulator; Glucokinase regulatory protein; Hexokinase 4 regulator; Hexokinase type IV; Hexokinase-4; Hexokinase-D; HK IV; HK4; OTTHUMP00000158533. Accession Number: Q14397. Expression Region: 1~625aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 70.7kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.