Learn More
Biomatik Corporation Recombinant Escherichia coli Chaperone protein DnaK (dnaK), 100UG
Supplier:  Biomatik Corporation RPC25379
Encompass_Preferred
Recombinant Escherichia coli Chaperone protein DnaK (dnaK) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Escherichia coli (strain UTI89 / UPEC). Target Name: dnaK. Target Synonyms: HSP70 Heat shock 70 kDa protein Heat shock protein 70. Accession Number: Q1RGI8. Expression Region: 1~638aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 71.1kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5423