Learn More
Biomatik Corporation Recombinant Arabidopsis thaliana Histone deacetylase HDT2 (HDT2), 100UG
Supplier:  Biomatik Corporation RPC25398
Encompass_Preferred
Recombinant Arabidopsis thaliana Histone deacetylase HDT2 (HDT2) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Arabidopsis thaliana (Mouse-ear cress). Target Name: HDT2. Target Synonyms: HD-tuins protein 2Histone deacetylase 2b. Accession Number: Q56WH4. Expression Region: 1~306aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 34.3kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5425