Learn More
Biomatik Corporation Recombinant Mouse Lymphocyte antigen 6E (Ly6e), 100UG
Supplier: Biomatik Corporation RPC25405
Recombinant Mouse Lymphocyte antigen 6E (Ly6e) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Mouse (Mus musculus). Target Name: Ly6e. Target Synonyms: Stem cell antigen 2Thymic shared antigen 1TSA-1. Accession Number: Q64253. Expression Region: 27~108aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 24.8kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.