Learn More
Biomatik Corporation Recombinant Staphylococcus aureus Gamma-hemolysin component C (hlgC), 100UG
Supplier:  Biomatik Corporation RPC25425
Encompass_Preferred
Recombinant Staphylococcus aureus Gamma-hemolysin component C (hlgC) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Staphylococcus aureus (strain N315). Target Name: hlgC. Target Synonyms: hlgC; SA2208Gamma-hemolysin component C. Accession Number: Q7A3S2. Expression Region: 30~315aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 34.6kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5430