Learn More
Biomatik Corporation Recombinant Cricetulus griseus Protein disulfide-isomerase (P4HB), 100UG
Supplier:  Biomatik Corporation RPC25466
Encompass_Preferred
Recombinant Cricetulus griseus Protein disulfide-isomerase (P4HB) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Chinese hamster. Target Name: P4HB. Target Synonyms: Prolyl 4-hydroxylase subunit betap58. Accession Number: Q8R4U2. Expression Region: 20~509aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 57kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5435