Learn More
Biomatik Corporation Recombinant Human Dedicator of cytokinesis protein 8 (DOCK8), partial, 100UG
Supplier:  Biomatik Corporation RPC25478
Encompass_Preferred
Recombinant Human Dedicator of cytokinesis protein 8 (DOCK8), partial is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: DOCK8. Target Synonyms: DOCK8Dedicator of cytokinesis protein 8. Accession Number: Q8NF50. Expression Region: 560~729aa. Tag Info: N-Terminal 10Xhis-Tagged. Theoretical MW: 21.8kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5438