Learn More
Biomatik Corporation Recombinant Mouse Plexin domain-containing protein 1 (Plxdc1), partial, 100UG
Supplier:  Biomatik Corporation RPC25480
Encompass_Preferred
Recombinant Mouse Plexin domain-containing protein 1 (Plxdc1), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Mouse (Mus musculus). Target Name: Plxdc1. Target Synonyms: Tumor endothelial marker 7. Accession Number: Q91ZV7. Expression Region: 20~426aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 47.3kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5439