Learn More
Biomatik Corporation Recombinant Macaca fascicularis T-cell surface glycoprotein CD3 gamma chain (CD3G), partial, 100UG
Supplier:  Biomatik Corporation RPC25493
Encompass_Preferred
Recombinant Macaca fascicularis T-cell surface glycoprotein CD3 gamma chain (CD3G), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Macaca fascicularis (Crab-eating macaque) (Cynomolgus monkey). Target Name: CD3G. Target Synonyms: T-cell receptor T3 gamma chainCD_antigen: CD3g. Accession Number: Q95LI7. Expression Region: 23~113aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 12.5kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5440