Learn More
Biomatik Corporation Recombinant Mouse 14-3-3 protein beta/alpha (Ywhab), 100UG
Supplier:  Biomatik Corporation RPC25537
Encompass_Preferred
Recombinant Mouse 14-3-3 protein beta/alpha (Ywhab) is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Mouse (Mus musculus). Target Name: Ywhab. Target Synonyms: Protein kinase C inhibitor protein 1. Accession Number: Q9CQV8. Expression Region: 1~246aa. Tag Info: N-Terminal 10Xhis-Tagged. Theoretical MW: 30.6kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5445