Learn More
Biomatik Corporation Recombinant Human Protein S100-A14 (S100A14), 100UG
Catalog No. 501505446
Encompass_Preferred
Recombinant Human Protein S100-A14 (S100A14) is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: S100A14. Target Synonyms: S100 calcium-binding protein A14S114S100A15. Accession Number: Q9HCY8. Expression Region: 1~104aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 13.7kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5446 Supplier Biomatik Corporation Supplier No. RPC25539
Recombinant Human Protein S100-A14 (S100A14) is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: S100A14. Target Synonyms: S100 calcium-binding protein A14S114S100A15. Accession Number: Q9HCY8. Expression Region: 1~104aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 13.7kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.