Learn More
Biomatik Corporation Recombinant Human Acetyl-coenzyme A synthetase 2-like, mitochondrial (ACSS1), 100UG
Supplier:  Biomatik Corporation RPC25546
Encompass_Preferred
Recombinant Human Acetyl-coenzyme A synthetase 2-like, mitochondrial (ACSS1) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: ACSS1. Target Synonyms: Acetate--CoA ligase 2Acetyl-CoA synthetase 2. Accession Number: Q9NUB1. Expression Region: 38~689aa. Tag Info: N-Terminal 10Xhis-Tagged. Theoretical MW: 73.6kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5447