Learn More
Biomatik Corporation Recombinant human Multiple inositol polyphosphate phosphatase 1 (MINPP1), 100UG
Supplier:  Biomatik Corporation RPC25569
Encompass_Preferred
Recombinant human Multiple inositol polyphosphate phosphatase 1 (MINPP1) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: MINPP1. Target Synonyms: 2,3-bisphosphoglycerate 3-phosphatase (EC:3.1.3.80); 2,3-BPG phosphataseInositol (1,3,4,5)-tetrakisphosphate 3-phosphatase; Ins(1,3,4,5)P(4) 3-phosphatase. Accession Number: Q9UNW1. Expression Region: 31~487aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 54.1kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5448