Learn More
Biomatik Corporation Recombinant A Disintegrin And Metalloproteinase With Thrombospondin 7 (ADAMTS7), 100UG
Supplier:  Biomatik Corporation RPU40032
Encompass_Preferred
Recombinant Human A Disintegrin And Metalloproteinase With Thrombospondin 7 (ADAMTS7) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: A Disintegrin And Metalloproteinase With Thrombospondin 7 (ADAMTS7). Target Synonyms: COMPase. Accession Number: Q9UKP4. Gene ID: 11173. Expression Region: Val1589~Arg1686. Tag Info: N-terminal His Tag. Theoretical MW: 15kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5450