Learn More
Biomatik Corporation Recombinant Adrenomedullin (ADM), 100UG
Supplier:  Biomatik Corporation RPU40122
Encompass_Preferred
Recombinant Mouse Adrenomedullin (ADM) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Adrenomedullin (ADM). Target Synonyms: AM; PAMP; Proadrenomedullin N-20 terminal peptide; ProAM N-terminal 20 peptide. Accession Number: P97297. Gene ID: 11535. Expression Region: Pro24~Ala171. Tag Info: N-terminal His Tag. Theoretical MW: 22kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5452