Learn More
Biomatik Corporation Recombinant Alanine Aminotransferase (ALT), 100UG
Supplier:  Biomatik Corporation RPU40146
Encompass_Preferred
Recombinant Human Alanine Aminotransferase (ALT) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Alanine Aminotransferase (ALT). Target Synonyms: GPT; ALAT; SGPT; AAT1; GPT1; Serum Glutamic Pyruvic Transaminase; Alanine Transaminase; Glutamate pyruvate transaminase 1; Glutamic--alanine transaminase 1. Accession Number: P24298. Gene ID: 2875. Expression Region: Ile255~Thr492. Tag Info: N-terminal His Tag. Theoretical MW: 27kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5454