Learn More
Biomatik Corporation Recombinant Alpha-1-Acid Glycoprotein (a1AGP), 100UG
Supplier:  Biomatik Corporation RPU40164
Encompass_Preferred
Recombinant Human Alpha-1-Acid Glycoprotein (a1AGP) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Alpha-1-Acid Glycoprotein (a1AGP). Target Synonyms: AGP1; a1-AGP; ORM1; OMD 1; Orosomucoid 1. Accession Number: P02763. Gene ID: 5004. Expression Region: Gln19~Ser201. Tag Info: N-terminal His Tag. Theoretical MW: 23kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5455