Learn More
Biomatik Corporation Recombinant Alpha-1-Microglobulin/Bikunin Precursor (a1M), 100UG
Supplier:  Biomatik Corporation RPU40179
Encompass_Preferred
Recombinant Mouse Alpha-1-Microglobulin (a1M) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Alpha-1-Microglobulin (a1M). Target Synonyms: AMBP; UTI; HCP; EDC1; HI30; IATIL; ITILC; ITI; ITIL; Alpha 1 Microglobulin/Bikunin Precursor; Growth-inhibiting protein 19; Uristatin; Uronic-Acid-Rich Protein; Trypstatin. Accession Number: Q07456. Gene ID: 11699. Expression Region: Arg18~Ala202. Tag Info: N-terminal His Tag. Theoretical MW: 24kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5456