Learn More
Biomatik Corporation Recombinant Alpha-2-Glycoprotein 1, Zinc Binding (aZGP1), 100UG
Supplier:  Biomatik Corporation RPU40184
Encompass_Preferred
Recombinant Mouse Alpha-2-Glycoprotein 1, Zinc Binding (aZGP1) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Alpha-2-Glycoprotein 1, Zinc Binding (aZGP1). Target Synonyms: ZA2G; ZAG. Accession Number: Q64726. Gene ID: 12007. Expression Region: Glu178~Gln307. Tag Info: N-terminal His Tag. Theoretical MW: 17kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5457