Learn More
Biomatik Corporation Recombinant Rabies virus Glycoprotein (G), partial, 100UG
Catalog No. 501504622
Encompass_Preferred
Recombinant Rabies virus Glycoprotein (G), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Rabies virus (strain PM) (RABV). Target Name: G. Target Synonyms: GGlycoprotein. Accession Number: A3RM22. Expression Region: 20~459aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 65.4kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4622 Supplier Biomatik Corporation Supplier No. RPC20143
Recombinant Rabies virus Glycoprotein (G), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Rabies virus (strain PM) (RABV). Target Name: G. Target Synonyms: GGlycoprotein. Accession Number: A3RM22. Expression Region: 20~459aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 65.4kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.