Learn More
Biomatik Corporation Recombinant Alpha-Fodrin (SPTAN1), 100UG
Catalog No. 501505459
Encompass_Preferred
Recombinant Human Alpha-Fodrin (SPTAN1) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Alpha-Fodrin (SPTAN1). Target Synonyms: SPTAN1; NEAS; (ALPHA)II-SPECTRIN; Spectrin Alpha Non-Erythrocytic 1; Spectrin Alpha Chain,Brain; Spectrin, non-erythroid alpha subunit. Accession Number: Q13813. Gene ID: 6709. Expression Region: Ser289~Gly408. Tag Info: N-terminal His Tag. Theoretical MW: 15kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5459 Supplier Biomatik Corporation Supplier No. RPU40211
Recombinant Human Alpha-Fodrin (SPTAN1) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Alpha-Fodrin (SPTAN1). Target Synonyms: SPTAN1; NEAS; (ALPHA)II-SPECTRIN; Spectrin Alpha Non-Erythrocytic 1; Spectrin Alpha Chain,Brain; Spectrin, non-erythroid alpha subunit. Accession Number: Q13813. Gene ID: 6709. Expression Region: Ser289~Gly408. Tag Info: N-terminal His Tag. Theoretical MW: 15kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.